N-alpha-Boc-L-tryptophanol 98%

Lieferant: Apollo Scientific

Synonyme: (S)-2-(tert-Butoxycarbonyl)amino-3-(3-indolyl)propanol, N-ɑ-(tert-Butoxycarbonyl)-L-tryptophanol, (S)-2-(Boc-amino)-3-(3-indolyl)propanol, Boc-Tryptophanol, N-Boc-L-tryptophanol

OR316000-25G
APOSOR316000-25GEA 145 EUR
APOSOR316000-25G
N-alpha-Boc-L-tryptophanol 98%
N-alpha-Boc-L-tryptophanol
Formel: C₁₆H₂₂N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00235953
CAS-Nummer: 82689-19-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR