N-Boc-L-cyclohexylglycinol

Lieferant: Apollo Scientific

Synonyme: (S)-tert-Butyl (1-cyclohexyl-2-hydroxyethyl)carbamate

OR316005-5G
APOSOR316005-5GEA 165 EUR
APOSOR316005-5G
N-Boc-L-cyclohexylglycinol
N-Boc-L-cyclohexylglycinol
Formel: C₁₃H₂₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04112591
CAS-Nummer: 107202-39-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR