(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-tert-Butyl 2-Methyl (2S,4S)-4-Hydroxypyrrolidine-1,2-dicarboxylate, (2S,4S)-1-tert-Butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, (2S,4S)-4-Hydroxypyrrolidine-1,2-dicarboxylic Acid 1-tert-Butyl 2-Methyl Ester, N-Boc-cis-4-hydroxy-L-proline Methyl Ester, Boc-cis-Hyp-OMe

OR316007-100G
APOSOR316007-100GEA 116 EUR
APOSOR316007-100G
(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate
(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate
Formel: C₁₁H₁₉NO₅
Molecular Weight: 245.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00237541
CAS-Nummer: 102195-79-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR