(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol

Lieferant: Apollo Scientific
Achtung

Synonyme: Boc-D-Phenylglycinol, (-)-N-Boc-D-α-phenylglycinol

OR316012-500G
APOSOR316012-500GEA 310 EUR
APOSOR316012-500G
(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol
(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol
Formel: C₁₃H₁₉NO₃
Molecular Weight: 237,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00274205
CAS-Nummer: 102089-74-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR