Boc-D-valinol

Lieferant: Apollo Scientific
Achtung

Synonyme: N-BOC-D-Valinol, Boc-D-Valinol

OR316019-5G
APOSOR316019-5GEA 24.8 EUR
APOSOR316019-5G
Boc-D-valinol
Boc-D-valinol
Formel: C₁₀H₂₁NO₃
Siedepunkt: 218 °C
Dichte: 0,995 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00235960
CAS-Nummer: 106391-87-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR