5-Isopropyl-1,3-cyclohexandion

Lieferant: Apollo Scientific

Synonyme: 5-Isopropylcyclohexane-1,3-dione

OR316026-1G
APOSOR316026-1GEA 85.7 EUR
APOSOR316026-1G
5-Isopropyl-1,3-cyclohexandion
5-Isopropyl-1,3-cyclohexandion
Formel: C₉H₁₄O₂·H₂O
Molecular Weight: 154.21 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00075263
CAS-Nummer: 18456-87-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR