D-allo-Threonin

Lieferant: Apollo Scientific
Achtung

Synonyme: H-D-Allo-Thr-OH, (2R,3R)-2-Amino-3-hydroxybutanoic acid

OR316027-25G
APOSOR316027-25GEA 119 EUR
APOSOR316027-25G
D-allo-Threonin
D-allo-Threonin
Formel: C₄H₉NO₃
Molecular Weight: 119.12 g/mol
Schmelzpunkt: 268…276 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00004526
CAS-Nummer: 24830-94-2
EINECS: 246-488-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR