Ethyl-2-oxocyclopentaneacetate

Lieferant: Apollo Scientific

OR316028-5G
APOSOR316028-5GEA 82.8 EUR
APOSOR316028-5G
Ethyl-2-oxocyclopentaneacetate
Ethyl-2-oxocyclopentaneacetate
Formel: C₉H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00044715
CAS-Nummer: 20826-94-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR