D-tert-Leucin (H-D-Tle-OH) 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: H-D-Tle-OH

OR316030-25G
APOSOR316030-25GEA 82.8 EUR
APOSOR316030-25G
D-tert-Leucin (H-D-Tle-OH) 97%
D-tert-Leucin (H-D-Tle-OH)
Formel: C₆H₁₃NO₂
Molecular Weight: 131.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004265
CAS-Nummer: 26782-71-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR