N-Boc-L-phenylalaninol

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-(-)-BOC-Phenylalaninol, Boc-Phenylalaninol, (S)-tert-butyl 1-hydroxy-3-phenylpropan-2-ylcarbamate

OR316043-100G
APOSOR316043-100GEA 69.6 EUR
APOSOR316043-100G
N-Boc-L-phenylalaninol
N-Boc-L-phenylalaninol
Formel: C₁₄H₂₁NO₃
Molecular Weight: 251,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00076976
CAS-Nummer: 66605-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR