(2R,5S)-2-Trichloromethyl-3-oxa-1-azabicyclo[3.3.0]octan-4-one

Lieferant: Apollo Scientific

OR316050-25G
APOSOR316050-25GEA 98.6 EUR
APOSOR316050-25G
(2R,5S)-2-Trichloromethyl-3-oxa-1-azabicyclo[3.3.0]octan-4-one
(2R,5S)-2-Trichloromethyl-3-oxa-1-azabicyclo[3.3.0]octan-4-one
Formel: C₇H₈Cl₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08274464
CAS-Nummer: 97538-67-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR