tert-Butyl (R)-(-)-2-Hydroxy-3,3-Dimethylbutyrate

Lieferant: Apollo Scientific
Achtung

OR316052-1G
APOSOR316052-1GEA 2660 EUR
APOSOR316052-1G
tert-Butyl (R)-(-)-2-Hydroxy-3,3-Dimethylbutyrate
tert-Butyl (R)-(-)-2-Hydroxy-3,3-Dimethylbutyrate
Formel: C₁₀H₂₀O₃
Molecular Weight: 188,264 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16657093
CAS-Nummer: 1349685-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR