4-(tert-Butoxy)benzeneboronic acid, pinacol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-tert-Butoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(tert-Butoxy)phenylboronic Acid Pinacol Ester

OR3161-1G OR3161-250MG
APOSOR3161-1GEA 33.5 EUR
APOSOR3161-1G APOSOR3161-250MG
4-(tert-Butoxy)benzeneboronic acid, pinacol ester
4-(tert-Butoxy)benzeneboronic acid, pinacol ester
Formel: C₁₆H₂₅BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08741438
CAS-Nummer: 938063-51-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR