6-Bromo-3-iodo-1H-pyrrolo[3,2-c]pyridine 99%

Lieferant: Apollo Scientific
Achtung

OR31655-1G OR31655-250MG
APOSOR31655-1GEA 325 EUR
APOSOR31655-1G APOSOR31655-250MG
6-Bromo-3-iodo-1H-pyrrolo[3,2-c]pyridine 99%
6-Bromo-3-iodo-1H-pyrrolo[3,2-c]pyridine
Formel: C₇H₄BrIN₂
Lagertemperatur: Kühlschrank
MDL: MFCD09749987
CAS-Nummer: 1000341-73-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR