6-Bromo-1H-pyrrolo[3,2-c]pyridine 99%

Lieferant: Apollo Scientific
Achtung

OR31656-250MG
APOSOR31656-250MGEA 36.4 EUR
APOSOR31656-250MG
6-Bromo-1H-pyrrolo[3,2-c]pyridine 99%
6-Bromo-1H-pyrrolo[3,2-c]pyridine
Formel: C₇H₅BrN₂
Molecular Weight: 197,0320 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08690134
CAS-Nummer: 1000342-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR