4-Brom-3-methylbenzamid
Lieferant: Apollo Scientific
|
Achtung
|
OR3168-25G
APOSOR3168-25GEA
91.5
EUR
APOSOR3168-25G
4-Brom-3-methylbenzamid
4-Brom-3-methylbenzamid
Formel:
C₈H₈BrNO Molecular Weight: 214.06 g/mol Schmelzpunkt: 177…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672914 CAS-Nummer: 170229-98-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...