2-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine

Lieferant: Apollo Scientific

OR317001-1G
APOSOR317001-1GEA 116 EUR
APOSOR317001-1G
2-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
2-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
Formel: C₆H₃ClIN₃
Molecular Weight: 279,4700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189363
CAS-Nummer: 1060815-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR