Methyl 7H-pyrrolo[2,3-d]pyrimidine-4-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR317002-1G
APOSOR317002-1GEA
224
EUR
APOSOR317002-1G
Methyl 7H-pyrrolo[2,3-d]pyrimidine-4-carboxylate
Methyl 7H-pyrrolo[2,3-d]pyrimidine-4-carboxylate
Formel:
C₈H₇N₃O₂ Molecular Weight: 177,16 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD17015873 CAS-Nummer: 1095822-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...