4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylic acid

Lieferant: Apollo Scientific

OR317006-1G
APOSOR317006-1GEA 119 EUR
APOSOR317006-1G
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylic acid
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylic acid
Formel: C₇H₄ClN₃O₂
Molecular Weight: 197,5800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11518908
CAS-Nummer: 186519-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR