5-Bromo-7H-pyrrolo[2,3-d]pyrimidine

Lieferant: Apollo Scientific
Achtung

OR317007-5G
APOSOR317007-5GEA 149 EUR
APOSOR317007-5G
5-Bromo-7H-pyrrolo[2,3-d]pyrimidine
5-Bromo-7H-pyrrolo[2,3-d]pyrimidine
Formel: C₆H₄BrN₃
Molecular Weight: 198.0200 g/mol
Schmelzpunkt: 199…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01877574
CAS-Nummer: 175791-49-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR