2,4-Dichloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine

Lieferant: Apollo Scientific
Achtung

OR317008-1G
APOSOR317008-1GEA 21.9 EUR
APOSOR317008-1G
2,4-Dichloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
2,4-Dichloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
Formel: C₆H₂Cl₂IN₃
Molecular Weight: 313,911 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13189364
CAS-Nummer: 1012785-51-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR