2-Chloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine

Lieferant: Apollo Scientific

OR317013-1G
APOSOR317013-1GEA 221 EUR
APOSOR317013-1G
2-Chloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine
2-Chloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine
Formel: C₇H₆ClN₃
Lagertemperatur: Kühlschrank
MDL: MFCD13189420
CAS-Nummer: 1060816-67-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR