6-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
OR317016-1G
APOSOR317016-1GEA
217
EUR
APOSOR317016-1G
6-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
6-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
Formel:
C₆H₃BrClN₃ Schmelzpunkt: 258 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08276174 CAS-Nummer: 784150-41-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...