2,4-Dichloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine
Lieferant: Apollo Scientific
OR317022-1G
APOSOR317022-1GEA
230
EUR
APOSOR317022-1G
2,4-Dichloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine
2,4-Dichloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine
Formel:
C₇H₅Cl₂N₃ Molecular Weight: 202,0407 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13193622 CAS-Nummer: 1192711-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...