6-(p-Toluenesulphonyl)-2-oxa-6-azaspiro[3.3]heptane
Lieferant: Apollo Scientific
OR317036-5G
APOSOR317036-5GEA
0
EUR
APOSOR317036-5G
6-(p-Toluenesulphonyl)-2-oxa-6-azaspiro[3.3]heptane
6-(p-Toluenesulphonyl)-2-oxa-6-azaspiro[3.3]heptane
Formel:
C₁₂H₁₅NO₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD13180735 CAS-Nummer: 13573-28-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...