3-Amino-oxetane-3-carboxylic acid methyl ester

Lieferant: Apollo Scientific

Synonyme: Methyl 3-aminooxetane-3-carboxylate

OR317038-1G
APOSOR317038-1GEA 322 EUR
APOSOR317038-1G
3-Amino-oxetane-3-carboxylic acid methyl ester
3-Amino-oxetane-3-carboxylic acid methyl ester
Formel: C₅H₉NO₃
Molecular Weight: 131,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20922764
CAS-Nummer: 1363383-31-6
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR