Toluene-4-sulphonic acid oxetan-2-yl methyl ester
Lieferant: Apollo Scientific
Synonyme:
Oxetan-2-ylmethyl 4-methylbenzenesulphonate
OR317041-1G
APOSOR317041-1GEA
184
EUR
APOSOR317041-1G
Toluene-4-sulphonic acid oxetan-2-yl methyl ester
Toluene-4-sulphonic acid oxetan-2-yl methyl ester
Formel:
C₁₁H₁₄O₄S Molecular Weight: 242,291 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08544404 CAS-Nummer: 115845-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...