1-Oxa-6-azaspiro[3.5]nonane oxalate

Lieferant: Apollo Scientific

Synonyme: 1-Oxa-6-azaspiro[3.5]nonane hemioxalate

OR317043-1G
APOSOR317043-1GEA 453 EUR
APOSOR317043-1G
1-Oxa-6-azaspiro[3.5]nonane oxalate
1-Oxa-6-azaspiro[3.5]nonane oxalate
Formel: C₁₆H₂₈N₂O₆
Molecular Weight: 344,41 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27923648
CAS-Nummer: 1523606-44-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR