2-Oxa-5-azaspiro[3.4]octane oxalate 96%

Lieferant: Apollo Scientific
Achtung

OR317053-250MG OR317053-500MG
APOSOR317053-250MGEA 0 EUR
APOSOR317053-250MG APOSOR317053-500MG
2-Oxa-5-azaspiro[3.4]octane oxalate 96%
2-Oxa-5-azaspiro[3.4]octane oxalate
Formel: C₆H₁₁NO·C₂H₂O₄
Molecular Weight: 203.19 g/mol
Schmelzpunkt: 85…94 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD18325162
CAS-Nummer: 1389264-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR