cis-3-Amino-cyclobutanecarboxylic acid hydrochloride

Lieferant: Apollo Scientific

OR317054-1G
APOSOR317054-1GEA 0 EUR
APOSOR317054-1G
cis-3-Amino-cyclobutanecarboxylic acid hydrochloride
cis-3-Amino-cyclobutanecarboxylic acid hydrochloride
Formel: C₅H₉NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD09971691
CAS-Nummer: 84182-59-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR