Diisopropyl-3,3-dimethoxycyclobutane-1,1-dicarboxylate

Lieferant: Apollo Scientific

Synonyme: Diisopropyl 3,3-dimethoxycyclobutane-1,1-dicarboxylate

OR317057-5G
APOSOR317057-5GEA 0 EUR
APOSOR317057-5G
Diisopropyl-3,3-dimethoxycyclobutane-1,1-dicarboxylate
Diisopropyl-3,3-dimethoxycyclobutane-1,1-dicarboxylate
Formel: C₁₄H₂₄O₆
Molecular Weight: 288,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06742779
CAS-Nummer: 115118-68-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR