Methyl trans-3-Amino-cyclobutanecarboxylate hydrochloride
Lieferant: Apollo Scientific
Synonyme:
trans-Methyl 3-aminocyclobutanecarboxylate hydrochloride
OR317065-1G
APOSOR317065-1GEA
114
EUR
APOSOR317065-1G
Methyl trans-3-Amino-cyclobutanecarboxylate hydrochloride
Methyl trans-3-Amino-cyclobutanecarboxylate hydrochloride
Formel:
C₆H₁₁NO₂·ClH Molecular Weight: 165.618 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17977174 CAS-Nummer: 74316-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...