2-Cyclobutylideneacetic acid

Lieferant: Apollo Scientific
Achtung

OR317078-1G
APOSOR317078-1GEA 0 EUR
APOSOR317078-1G
2-Cyclobutylideneacetic acid
2-Cyclobutylideneacetic acid
Formel: C₆H₈O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09865731
CAS-Nummer: 25021-04-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR