3-(Boc-aminomethyl)cyclobutanone

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl N-[(3-oxocyclobutyl)methyl]carbamate, tert-Butyl ((3-oxocyclobutyl)methyl)carbamate

OR317084-1G
APOSOR317084-1GEA 271 EUR
APOSOR317084-1G
3-(Boc-aminomethyl)cyclobutanone
3-(Boc-aminomethyl)cyclobutanone
Formel: C₁₀H₁₇NO₃
Molecular Weight: 199,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11499004
CAS-Nummer: 130369-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR