tert-Butyl-7-azaspiro[3.5]nonan-2-ylcarbamate 97%

Lieferant: Apollo Scientific

Synonyme: tert-butyl N-{7-azaspiro[3.5]nonan-2-yl}carbamate, tert-Butyl 7-azaspiro[3.5]nonan-2-ylcarbamate, 2-(Boc-amino)-7-azaspiro[3.5]nonane

OR317096-1G
APOSOR317096-1GEA 393 EUR
APOSOR317096-1G
tert-Butyl-7-azaspiro[3.5]nonan-2-ylcarbamate 97%
tert-Butyl-7-azaspiro[3.5]nonan-2-ylcarbamate
Formel: C₁₃H₂₄N₂O₂
Molecular Weight: 240,3400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12198553
CAS-Nummer: 147611-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR