Methyl cis-3-hydroxycyclobutanecarboxylate

Lieferant: Apollo Scientific

Synonyme: cis-Methyl 3-hydroxycyclobutanecarboxylate

OR317100-1G
APOSOR317100-1GEA 85.7 EUR
APOSOR317100-1G
Methyl cis-3-hydroxycyclobutanecarboxylate
Methyl cis-3-hydroxycyclobutanecarboxylate
Formel: C₆H₁₀O₃
Molecular Weight: 130,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18483124
CAS-Nummer: 63485-50-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR