3,3-Dimethoxycyclobutane-1-carboxylate methyl ester 96%

Lieferant: Apollo Scientific

Synonyme: Methyl 3,3-dimethoxycyclobutanecarboxylate, methyl 3,3-dimethoxycyclobutane-1-carboxylate

OR317106-25G
APOSOR317106-25GEA 63.8 EUR
APOSOR317106-25G
3,3-Dimethoxycyclobutane-1-carboxylate methyl ester 96%
3,3-Dimethoxycyclobutane-1-carboxylate methyl ester
Formel: C₈H₁₄O₄
Molecular Weight: 174.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11111944
CAS-Nummer: 98231-07-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR