[1-(Aminomethyl)cyclobutyl]methanol

Lieferant: Apollo Scientific

Synonyme: (1-(Aminomethyl)cyclobutyl)methanol

OR317108-1G
APOSOR317108-1GEA 255 EUR
APOSOR317108-1G
[1-(Aminomethyl)cyclobutyl]methanol
[1-(Aminomethyl)cyclobutyl]methanol
Formel: C₆H₁₃NO
Molecular Weight: 115,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864389
CAS-Nummer: 2041-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR