Methyl-3-oxo-1-methylcyclobutanecarboxylate

Lieferant: Apollo Scientific

Synonyme: Methyl 1-methyl-3-oxocyclobutanecarboxylate

OR317122-1G
APOSOR317122-1GEA 194 EUR
APOSOR317122-1G
Methyl-3-oxo-1-methylcyclobutanecarboxylate
Methyl-3-oxo-1-methylcyclobutanecarboxylate
Formel: C₇H₁₀O₃
Molecular Weight: 142,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23105986
CAS-Nummer: 1408075-88-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR