trans-3-Methoxycyclobutanamine hydrochloride

Lieferant: Apollo Scientific

OR317140-1G
APOSOR317140-1GEA 351 EUR
APOSOR317140-1G
trans-3-Methoxycyclobutanamine hydrochloride
trans-3-Methoxycyclobutanamine hydrochloride
Formel: C₅H₁₂ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD22415180
CAS-Nummer: 1408074-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR