6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid methyl ester

Lieferant: Apollo Scientific

Synonyme: Methyl 6-(Boc-amino)spiro[3.3]heptane-2-carboxylate, Methyl 6-((tert-butoxycarbonyl)amino)spiro[3.3]heptane-2-carboxylate

OR317141-1G
APOSOR317141-1GEA 109 EUR
APOSOR317141-1G
6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid methyl ester
6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid methyl ester
Formel: C₁₄H₂₃NO₄
Molecular Weight: 269,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18425636
CAS-Nummer: 170508-14-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR