(6-Hydroxyspiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-butyl N-{6-hydroxyspiro[3.3]heptan-2-yl}carbamate, 2-(Boc-amino)-6-hydroxyspiro[3.3]heptane, tert-Butyl (6-hydroxyspiro[3.3]heptan-2-yl)carbamate

OR317142-1G
APOSOR317142-1GEA 188 EUR
APOSOR317142-1G
(6-Hydroxyspiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester 97%
(6-Hydroxyspiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester
Formel: C₁₂H₂₁NO₃
Molecular Weight: 227.31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09702342
CAS-Nummer: 1000933-99-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR