3-Hydroxy-1-methylazetidine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methyl-3-azetidinol Hydrochloride, 1-Methylazetidin-3-ol hydrochloride

OR317154-1G
APOSOR317154-1GEA 56.7 EUR
APOSOR317154-1G
3-Hydroxy-1-methylazetidine hydrochloride
3-Hydroxy-1-methylazetidine hydrochloride
Formel: C₄H₉NO·ClH
Molecular Weight: 123,581 g/mol
Schmelzpunkt: 87…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16605978
CAS-Nummer: 26687-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR