tert-Butyl-3-[(methylsulphonyl)oxy]-1-azetanecarboxylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Boc-3-Methanesulphonyloxyazetidine

OR317155-5G
APOSOR317155-5GEA 21.9 EUR
APOSOR317155-5G
tert-Butyl-3-[(methylsulphonyl)oxy]-1-azetanecarboxylate 97%
tert-Butyl-3-[(methylsulphonyl)oxy]-1-azetanecarboxylate
Formel: C₉H₁₇NO₅S
Molecular Weight: 251,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05664832
CAS-Nummer: 141699-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR