1-Boc-3-(iodoethyl)azetidine

Lieferant: Apollo Scientific

OR317172-1G
APOSOR317172-1GEA 258 EUR
APOSOR317172-1G
1-Boc-3-(iodoethyl)azetidine
1-Boc-3-(iodoethyl)azetidine
Formel: C₁₀H₁₈INO₂
Molecular Weight: 311,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20126205
CAS-Nummer: 158602-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR