1-Boc-3-mercapto-azetidine
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 3-mercaptoazetidine-1-carboxylate
OR317173-1G
APOSOR317173-1GEA
81.2
EUR
APOSOR317173-1G
1-Boc-3-mercapto-azetidine
1-Boc-3-mercapto-azetidine
Formel:
C₈H₁₅NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861743 CAS-Nummer: 941585-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...