7-Oxo-2,6-diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 7-oxo-2,6-diazaspiro[3.4]octane-2-carboxylate, 2-Boc-7-oxo-2,6-diaza-spiro[3.4]octane-2-carboxylate
OR317185-1G
APOSOR317185-1GEA
68.3
EUR
APOSOR317185-1G
7-Oxo-2,6-diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester
7-Oxo-2,6-diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester
Formel:
C₁₁H₁₈N₂O₃ Molecular Weight: 226,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18325190 CAS-Nummer: 1234616-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...