5-Boc-2,5-diazaspiro[3.5]nonane oxalate 95%

Lieferant: Apollo Scientific

OR317197-250MG
APOSOR317197-250MGEA 609 EUR
APOSOR317197-250MG
5-Boc-2,5-diazaspiro[3.5]nonane oxalate 95%
5-Boc-2,5-diazaspiro[3.5]nonane oxalate
Formel: C₁₂H₂₂N₂O₂·C₂H₂O₄
Molecular Weight: 316,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22575224
CAS-Nummer: 1408076-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR