N-(Azetidin-3-yl)methanesulphonamide hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR317200-1G
APOSOR317200-1GEA
178
EUR
APOSOR317200-1G
N-(Azetidin-3-yl)methanesulphonamide hydrochloride
N-(Azetidin-3-yl)methanesulphonamide hydrochloride
Formel:
C₄H₁₀N₂O₂S·ClH Molecular Weight: 186,66 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD20926152 CAS-Nummer: 1239205-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...