5-Oxa-2-azaspiro[3.4]octane oxalate (2:1)
Lieferant: Apollo Scientific
OR317208-1G
APOSOR317208-1GEA
0
EUR
APOSOR317208-1G
5-Oxa-2-azaspiro[3.4]octane oxalate (2:1)
5-Oxa-2-azaspiro[3.4]octane oxalate
Formel:
C₁₄H₂₄N₂O₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD27988000 CAS-Nummer: 1523618-29-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...